Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Lens Set
Eisco lens sets are ideal in the classroom for doing optical experiments comparing focal lengths and convex vs. concave lenses.
$15.25
Specifications
For Use With (Application) | For doing optical experiments comparing focal lengths and convex vs. concave lenses |
---|
Catalog Number | Mfr. No. | Diameter (English) Lens | Diameter (Metric) Lens | Product Type | Lens Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Lens | Diameter (Metric) Lens | Product Type | Lens Material | Price | Quantity & Availability | ||
S96196
|
Eisco™
FSCI017 |
1.5 in. | 38 mm | Lens, Glass | Glass |
Pack of 6 for $15.25
|
|
||
Description
Three diameters for a variety of uses.
- With ground and polished faces and ground edges
- One each: of double convex, plano-convex, converging concave-convex, diverging concave-convex, double concave, plano-concave
- Each size is set of six
Glass Lens Sets
- S96196: 50mm diameter
- S96197: 38mm diameter
- S96198: 50 mm diameter
Specifications
For doing optical experiments comparing focal lengths and convex vs. concave lenses |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title