Learn More
Eisco™ Physics Kit: Mechanics System 2 Products for Science Education
Students can explore common experiments such as Boyle's Law, The Cartesian Diver, and Bernoulli's Principle.
Manufacturer:  Eisco™ EIRQ02
Catalog No. S13465
Add to cart
Includes: Universal Base with knobs 2, Expansion clamp with rod (4 prong) 2, Support rod, rounded ends, length 350mm 4, Mariotte's bottle 1, Tube for Mariotte's bottle, glass 1, Hare's apparatus 1, Silicone tubing, 2 meters 1, Plastic boss head clamps with knobs 2, Force pump 1, Rubber stopper, 1 hole for Mariotte's bottle 1, Ping-pong ball 1, Base for capillary tube and communicating vessels 1, Beaker 250 mL, plastic 2, Styrofoam ball 4, Pascal apparatus ball 1, Food color 1, Drying tube, U shaped 1, Venturi's tube 1, Syringe 5mL 1, Syringe 20mL 1, U-tube manometer with stand 1, U-tube manometer base 1, Communicating vessels 1, Capillary tubes 1, Funnel, 10cm, plastic 1, Silicone grease 1, Culture tube 13 x 100, glass with screw cap 1, Small stopper that fits in Mariotte's bottle spout 1, Ostwald Viscometer 1
Description
Description
  • Use to teach 14 different fluid mechanics activities
  • Includes Activity Guides with Student worksheets
  • Contains 36 components, including all tubes and vessels one needs to investigate core concepts in introductory fluid mechanics
  • Packed in Aluminum case with foam cut outs
Experiment Guide covering 14 activities
  • Mariotte's Bottle
  • The Force Pump
  • Discovering Boyle's Law Using a Syringe
  • Pascal's Ball Aparatus
  • Communicating Vessels
  • Finding the Density of a Liquid with a U-Tube
  • Open Tube Manometer
  • Hare's Apparatus
  • Cartesian Diver
  • Bernoulli's Principle
  • Venturia's Tube
  • The Ostwald Viscometer
  • Capillary Tube
Includes:
Universal Base with knobs 2, Expansion clamp with rod (4 prong) 2, Support rod, rounded ends, length 350mm 4, Mariotte's bottle 1, Tube for Mariotte's bottle, glass 1, Hare's apparatus 1, Silicone tubing, 2 meters 1, Plastic boss head clamps with knobs 2, Force pump 1, Rubber stopper, 1 hole for Mariotte's bottle 1, Ping-pong ball 1, Base for capillary tube and communicating vessels 1, Beaker 250 mL, plastic 2, Styrofoam ball 4, Pascal apparatus ball 1, Food color 1, Drying tube, U shaped 1, Venturi's tube 1, Syringe 5mL 1, Syringe 20mL 1, U-tube manometer with stand 1, U-tube manometer base 1, Communicating vessels 1, Capillary tubes 1, Funnel, 10cm, plastic 1, Silicone grease 1, Culture tube 13 x 100, glass with screw cap 1, Small stopper that fits in Mariotte's bottle spout 1, Ostwald Viscometer 1
Specifications
Specifications
Mechanics System 2 Physics Kit
Education