Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EKLF Monoclonal Antibody (7B2), Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 61233

Description
KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.Specifications
EKLF | |
Monoclonal | |
1 mg/mL | |
12mM tris, 60mM sodium citrate with 120mM NaCl, 30% glycerol and 0.035% sodium azide; pH 8 | |
P46099 | |
Klf1 | |
This EkLF antibody was raised against a full-length recombinant protein to mouse EkLF (kruppel-like factor 1 (erythroid)). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG3 |
ChIP Assay, Immunoprecipitation, Western Blot | |
7B2 | |
Unconjugated | |
Klf1 | |
CDAN4; EKLF; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; HBFQTL6; INLU; KLF1; Krueppel-like factor 1; Kruppel like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; Nan; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1 | |
Mouse | |
Protein G | |
RUO | |
16596 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title