Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EKLF Monoclonal Antibody (7B2), Active Motif™
SDP

Mouse Monoclonal Antibody

$121.20 - $369.66

Specifications

Antigen EKLF
Clone 7B2
Concentration 1 mg/mL
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3255
SDP
Encompass
View Documents
Active Motif
61234
10 μg
Each for $121.20
Add to Cart
 
50-199-3254
SDP
Encompass
View Documents
Active Motif
61233
100 μg
Each for $369.66
Add to Cart
 
Description

Description

KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.
Specifications

Specifications

EKLF
1 mg/mL
Monoclonal
Liquid
RUO
12mM tris, 60mM sodium citrate with 120mM NaCl, 30% glycerol and 0.035% sodium azide; pH 8
CDAN4; EKLF; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; HBFQTL6; INLU; KLF1; Krueppel-like factor 1; Kruppel like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; Nan; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1
Klf1
IgG3
Protein G
Antibody
7B2
ChIP Assay, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Mouse
P46099
16596
This EkLF antibody was raised against a full-length recombinant protein to mouse EkLF (kruppel-like factor 1 (erythroid)).
Primary
-20°C, Avoid Freeze/Thaw Cycles
Klf1
SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit