Learn More
ELA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179431AP
Catalog No. 50-173-1891
Add to cart
Description
Description
The neutrophil form of elastase (EC 3.4.21.37) is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
Specifications
Specifications
ELA2
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08246, Q3UP87
Bone marrow serine protease, ELA2, ELANE, Elastase 2, elastase 2, neutrophil, GE, HLE, HNE, Human leukocyte elastase, Medullasin, NE, Neutrophil elastase, PMN E, PMN elastase
Rabbit
IgG
150 μL
-20°C
Primary
1991, 299606, 50701
Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
ELANE
Liquid
Elane
ELA2 Fusion Protein Ag12372
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat