Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELA3B Mouse anti-Human, Clone: CELA3B/1218, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14786

Catalog No. 89-034-869


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human ELA3B.

  • High specificity
  • High reactivity
Specifications

Specifications

ELA3B
CELA3B/1218
Mouse monoclonal antibody raised against partial recombinant human ELA3B.
In 10 mM PBS.
Liquid
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
ELISA (2-4 ug/mL for coating) Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P08861
ELA3B
Recombinant protein corresponding to amino acids 82-238 of human ELA3B.
Protein A/G purification
RUO
Primary
23436
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)