Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELAC1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ELAC1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155326
|
Novus Biologicals
NBP155326 |
100 μL |
Each of 1 for $436.00
|
|
Description
ELAC1 Polyclonal specifically detects ELAC1 in Human samples. It is validated for Western Blot.Specifications
ELAC1 | |
Polyclonal | |
Rabbit | |
Q9H777 | |
55520 | |
Synthetic peptides corresponding to ELAC1(elaC homolog 1 (E. coli)) The peptide sequence was selected from the N terminal of ELAC1. Peptide sequence MSMDVTFLGTGAAYPSPTRGASAVVLRCEGECWLFDCGEGTQTQLMKSQL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
D29elaC (E. coli) homolog 1, Deleted in Ma29, EC 3.1.26.11, elaC homolog 1 (E. coli), ElaC homolog protein 1, FLJ59261, Ribonuclease Z 1, RNase Z 1, tRNA 3 endonuclease 1, tRNA 3' processing endoribonuclease, tRNA Z (short form), tRNase Z 1, tRNase ZS, zinc phosphodiesterase ELAC protein 1 | |
ELAC1 | |
IgG | |
Affinity Purified | |
40 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title