Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elafin/Skalp Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159631AP

Catalog No. 50-173-0603


Add to cart

Description

Description

Neutrophil and pancreatic elastase-specific inhibitor of skin. It may prevent elastase-mediated tissue proteolysis.
Specifications

Specifications

Elafin/Skalp
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P19957
cementoin, Elafin, Elastase specific inhibitor, ESI, Peptidase inhibitor 3, PI 3, PI3, Protease inhibitor WAP3, SKALP, WAP3, WFDC14
Rabbit
IgG
150 μL
-20°C
Primary
5266
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PI3
Liquid
PI3
Elafin/Skalp Fusion Protein Ag8735
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit