Learn More
Alfa Aesar™ Elastase Inhibitor II
A potent irreversible inhibitor of human neutrophil elastase
Manufacturer:  Alfa Aesar™ J65641LB0
 View more versions of this product
Catalog No. AAJ65641LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
−30° to −10°C
A potent irreversible inhibitor of human neutrophil elastase
474.90g/mol
Moisture Sensitive
Signal Transduction Reagent
White
Solid
C20H31ClN4O7
HNE Inhibitor; MeOSuc-AAPA-CMK
Soluble in methanol or DMSO