Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastase Mouse anti-Human, Alexa Fluor™ 350, Clone: CELA3B/1374, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP254302AF350
Description
Elastase Monoclonal specifically detects Elastase in Human samples. It is validated for Western Blot, ELISA.Specifications
Elastase | |
CELA3B/1374 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
Mouse | |
IgG3 Kappa | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CELA3B | |
Recombinant human CELA3B protein fragment (aa82-238) (exact sequence is proprietary) | |
0.1 mL | |
Primary | |
23436 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title