Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase Mouse anti-Human, Alexa Fluor™ 532, Clone: CELA3B/1374, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254302AF532

Catalog No. NP24302A32


Add to cart

Description

Description

Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Flow Cytometry,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

Elastase
CELA3B/1374
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
Mouse
IgG3
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CELA3B
Recombinant human CELA3B protein fragment (aa82-238) (exact sequence is proprietary)
0.1 mL
Primary
23436
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit