Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase Mouse anti-Human, Clone: rCELA3B/1811, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30753710UG

 View more versions of this product

Catalog No. NB122547


Add to cart

Description

Description

Elastase Monoclonal specifically detects Elastase in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

Elastase
rCELA3B/1811
Unconjugated
10 mM PBS with 0.05% BSA
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
Recombinant human Elastase protein fragment (around aa 82-238) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Cancer, Neuroscience
Primary
23436
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit