Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastase Mouse anti-Human, Clone: rCELA3B/1811, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30753720UG
Description
Elastase Monoclonal specifically detects Elastase in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Elastase | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
Recombinant human Elastase protein fragment (around aa 82-238) (exact sequence is proprietary) | |
20 μg | |
Cancer, Neuroscience | |
23436 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
rCELA3B/1811 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction