Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase Mouse anti-Human, PE-Cyanine7, Clone: CELA3B/1811, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254489PECY7

Catalog No. N254489PEC7


Add to cart

Description

Description

Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Elastase
CELA3B/1811
Flow Cytometry
Conjugated, Purified
CELA3B
Recombinant human CELA3B protein fragment (aa82-238) (exact sequence is proprietary) produced in E.coli
Protein A or G purified
Cancer, Neuroscience
Primary
23436.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit