Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Elastase (Porcine), 2X
Used for Enzyme
Supplier: Abnova™ P5225
Description
#REF!Specifications
Enzyme Activity | |
Elastase (Porcine), 2x | |
25 mg | |
RUO | |
Elastase Assay; Isolation of Type II lung cells. | |
Pig | |
Native protein purified from Porcine Pancreas |
25.9kDa | |
Native protein purified from Porcine Pancreas | |
Store at 4°C. Do not freeze. This product is stable for at least 3 months. | |
ELASTASE | |
≥3 units per mg protein. One Unit cleaves one micromole of N-succinyl-L-alanyl-L-alanyl-L-alanine-p-nitroanilide per minute at 25°C, pH 8.0. | |
None | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction