Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase, porcine pancreas aqueous suspension

$182.40

Specifications

CAS 39445-21-1
Molecular Weight 25 kd
Synonym E.C. 3.4.21.36; Pancreatopeptidase E
Storage Requirements 2° to 8°C
Format Suspension
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ61753MC View Documents Alfa Aesar™
J61753MC
100mg Each for $182.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • A serine protease that acts on elastin
  • Suitable for the isolation of Type II lung cells
Specifications

Specifications

39445-21-1
E.C. 3.4.21.36; Pancreatopeptidase E
Suspension
Off-white to Pale Brown
25 kd
2° to 8°C
A serine protease that acts on elastin. Suitable for the isolation of Type II lung cells
Elastase
Safety and Handling

Safety and Handling

EINECSNumber : 254-453-6

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit