Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastase Rabbit anti-Human, Clone: CELA3B/2810R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Elastase
Clone CELA3B/2810R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122622
View Documents
Novus Biologicals
NBP30757420UG
20 μg
Each for $159.82
Add to cart
 
NB122621
View Documents
Novus Biologicals
NBP30757410UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Elastase
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E
Recombinant human Elastase protein fragment (around aa 82-238) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
CELA3B/2810R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Neuroscience
10 mM PBS with 0.05% BSA
23436
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit