Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin, 10% Solution, Spectrum™

E1217, 9007-58-3

Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner E12171KGBL

 View more versions of this product

Catalog No. 18-602-402


Add to cart

Description

Description

Spectrum™ Elastin, 10 Percent Solution , an elastic protein, allows body tissues to return to their shape after contracting or stretching. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
Specifications

Specifications

1 kg
Poly-Coated Glass Bottle
Elastin (10%), water (90%)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit