Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin Guinea Pig anti-Human, Polyclonal, Biologo Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$517.28 - $965.58

Specifications

Antigen Elastin
Concentration 1 mg/mL
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0755 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
EL25013
0.5 mL
Each for $965.58
Add to cart
 
50-210-0756 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
EL2501301
0.1 mL
Each for $517.28
Add to cart
 
Description

Description

Elastin Polyclonal antibody specifically detects Elastin in Human samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Specifications

Specifications

Elastin
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Unconjugated
Guinea Pig
Human
P15502
2006
IgG
Primary
-20°C
ELN
1 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative; pH
AI385707; AI480567; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; Eln; FLJ38671; FLJ43523; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS
Purified elastin from human skin
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit