Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elastin Rabbit anti-Human, Clone: ELN/3131R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30714810UG
Description
Elastin Monoclonal specifically detects Elastin in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Elastin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS | |
Recombinant full-length human Elastin protein | |
100 μg | |
Extracellular Matrix | |
2006 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
ELN/3131R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title