Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastin Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152571AP

Catalog No. 50-173-0098


Add to cart

Description

Description

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Elastin
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15502, P54320
elastin, ELN, SVAS, Tropoelastin, WBS, WS
Rabbit
IgG
150 μL
-20°C
Primary
13717, 2006
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
ELN
Liquid
ELN
Elastin Fusion Protein Ag7179
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit