Learn More
Fisherbrand™ Elbow with 3/32 in. ID - Natural Kynar - QC
SDP
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing, for a tube-to-tube connection.
Catalog No. 01-000-605
Add to cart
Description
Description
  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Compliance Certificates Available
Natural Kynar
Tube to Tube Connector for Medical, Pharmaceutical, Industrial applications
Out of direct sunlight, >20°F <100°F
Class 7 Cleanroom Manufactured
Elbow
100 Pack
Natural