Learn More
Cole-Parmer Masterflex Fitting, PVDF, Elbow, Hose Barb to Threaded Adapter, 3/8" ID x 1/2" NPT(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3070442
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-630-034