Learn More
Eisco™ Electric Clinostat Products for Science Education
For demonstration of heliotropism and geotropism.
Manufacturer:  Eisco™ BI0244
Catalog No. S24472
Add to cart
Description
Description
  • Germinating seedlings pinned to cork table rotated by electric motor at 4 RpH
  • Motor spindle-mounted and clamped for table to be set at any angle between horizontal and vertical
  • Acrylic cover of cork table shields seedlings from droughts
Specifications
Specifications
Education
Demonstration of heliotropism and geotropism
Acrylic cover shields seedlings from drought
100mm cork disc
Germinating seedlings pinned to cork table rotated by electro motor at 4 rph
Electric Clinostat