Learn More
Brady™ Electric Lockout Pouch Kit “Spanish”
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Electric Lockout Pouch Kit “Spanish” is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ 65042
Catalog No. 19-104-174
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Safety
Electrical risk lockouts
(1) lockout bag; (1) kd steel padlock, 2 in. shackle, blue; (1) no-hole circuit breaker, 277v; (1) multi-pole circuit breaker; (1) plug lockout, 110v; (1) lockout hasp; (2) lockout tags