Learn More
Brady™ Electrical Lockout Starter Kit, Latin America
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Electric Lockout Starter Kit is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ 121513
Catalog No. 19-104-168
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical risk lockouts
Peligro No Operar Tags
(1) lockout bag; (10) lockout tags; (2) 0.75 in. shackle steel padlocks, blue; (2) 2.0 in. shackle steel padlocks, blue; (1) plug lockout, 220/550v; (1) plug lockout, 110v; (1) wall switch lockout; (1) no-hole circuit breaker (pk 6); (1) multi-pole circuit breaker (pk 6); (3) safety hasps
18 in.
Safety
Yellow
9.75 in.
Canvas
18 in. x 9.75 in.