Learn More
Cole-Parmer Fluke T+PRO-1AC KIT Voltage and Continuity Tester  DFS Item
Manufacturer:  Cole-Parmer 2604644
The Fluke T + PRO is the ideal, full-featured tester for the demanding commercial and industrial electrician. The Fluke T + is the basic electrical tester for residential electricians and HVAC technicians. These testers include all of the advantages of a traditional solenoid tester, with added functionality, but none of the typical drawbacks. With voltage measurement, continuity, built-in flashlights, GFCI trip capability, and Fluke's legendary ruggedness and reliability, the tester is an excellent choice for most residential, commercial, and industrial applications.
  • Safer than traditional solenoid testers Available for sale in the United States and Canada only.
  • Catalog No. 50-630-999