Learn More
Bel-Art™ Electrode Holder
For multi-purpose flexible arm stand.
Supplier: Bel-Art™ F183171000
Description
Snaps onto the end of the Flexible Gooseneck Arm affording three stepped wells for holding an electrode, a reference electrode and an ATC probe.Specifications
Electrode Holder | |
Holding an electrode, a reference electrode and an ATC probe |