Learn More
Cole-Parmer Cole-Parmer High-Accuracy pH Probe, SJ/Glass/100Ohm RTD/10'; Tinned/BNC  DFS Item
Manufacturer:  Cole-Parmer 2701206
High-accuracy electrodes are perfect for testing where temperature variations make many pH electrodes unreliable. These single-junction electrodes feature an innovative design that stabilizes readings in less than 20 seconds, with an accuracy of ±0.02 pH! The pH value stays constant at any given temperature up to pH 13. Maximum pressure is 100 psi at 25°>C.
Catalog No. 50-633-688