Learn More
Cole-Parmer Cole-Parmer pH electrode, combination, rugged, glass body, interchangeble connection  DFS Item
Manufacturer:  Cole-Parmer 0565805
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
Catalog No. 50-633-508