Learn More
Cole-Parmer Cole-Parmer In-Line pH Electrode with Tuff-Tip, 100 Ohms ATC Element  DFS Item
Manufacturer:  Cole-Parmer 0599502
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.
Catalog No. 50-633-467