Learn More
Cole-Parmer Cole-Parmer Gel-Filled Side-Mount pH Probe, SJ/PVDF/100Ohm RTD; Tinned/BNC  DFS Item
Manufacturer:  Cole-Parmer 2701120
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.
Catalog No. 50-633-464