Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Electrode Storage Solution for Storing Glass and Combination pH Electrodes, Reagents
Electrode Storage Solution for Storing Glass and Combination pH Electrodes, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CE00160020F
Specifications
100°C | |
Electrode | |
20 L |
Colorless | |
Cubitainer | |
Completely miscible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction