Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ 1M KNO3 Electrolyte Solution

Features a special cap for easy refilling of electrodes. Mettler Toledo™ 1M KNO3 Electrolyte Solution provides a bridge electrolyte for optimum performance of DX series Ion-Selective Electrode (ISE) half-cells. It is used for determination of ammonium, potassium and nitrate.

Manufacturer:  Mettler Toledo™ 51350086

 View more versions of this product

Catalog No. 01-913-217


Add to cart

Description

Description

Measuring with ion-selective electrodes (ISE) is the easiest and most affordable way to determine ion concentration. However, ISEs require careful handling and the use of the correct solutions. It is important to fill the reference electrode of every ISE with the recommended electrolyte solution. The correct electrolyte will minimize junction potentials and provide optimum temperature and time response.

  • Recommended for use with DX-series ISE half-cells
  • Used for determination of ammonium, potassium and nitrate
  • Equipped with a special cap to facilitate easy filling of electrodes
  • The electrolyte must be refilled or replaced regularly for good electrode performance
  • Available in 25mL and 250mL bottles, and a 6 x 250mL bulk pack
Specifications

Specifications

6x 250 mL
ISE Bridge Electrolyte Solution
Colorless
Electrolyte 1 mol/L KNO3, 6 x 250 mL
6 x 250 mL
6
1 mol/L KNO3
Electrolyte
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit