Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrolyte Solution, 1mol/L KNO3, Mettler Toledo™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
125mL
250mL
500mL

Chemical Identifiers

7757-79-1
KNO3
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
  • Quality solutions for accurate results
  • Proper storage and care of pH and redox electrodes ensures a long life and accurate results

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 101.10
Physical Form Liquid
Quantity 250mL
Chemical Name or Material Electrolyte Solution, 1mol/L KNO3