Learn More
SI Analytics™ Electrolyte Solution
Supplied in a 250 mL DURAN™ Bottle
$38.85 - $73.50
Specifications
Packaging DURAN™ Bottle (250 mL)
Filling Solution Type Electrolyte Solution
View More Specs
Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
15-177-897
View Documents
SI Analytics™
285138324
Acetic Acid Saturated with Lithium Chloride Each for $73.50
Add to cart
15-177-895
View Documents
SI Analytics™
285138427
3 M KCl Each for $46.35
Add to cart
15-177-896
View Documents
SI Analytics™
285138349
2 M KNO3 and 0.001 M KCl Each for $38.85
Add to cart
Specifications
Specifications
DURAN™ Bottle (250 mL)
Electrolyte Solution