Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrolyte Solution, 3mol/L KCl, AgCl Saturated, Mettler Toledo™

Click to view available options
Quantity:
125mL
250mL
7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
  • Quality solutions for accurate results
  • Proper storage and care of pH and redox electrodes ensures a long life and accurate results
Physical Form Liquid
Quantity 125mL
Chemical Name or Material Electrolyte Solution, 3mol/L KCl, AgCl Saturated