Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrolyte Solution, 3mol/L KCl, Mettler Toledo™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
250mL
500mL

Chemical Identifiers

7447-40-7
ClK
74.548
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Potassium Chloride
4873
CHEBI:32588
  • Quality solutions for accurate results
  • Proper storage and care of pH and redox electrodes ensures a long life and accurate results

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

CAS 7447-40-7
Molecular Formula ClK
Synonym Potassium Chloride
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium;chloride
Molecular Weight (g/mol) 74.548
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.55
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Electrolyte Solution, 3mol/L KCl
Show More Show Less