Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Electronic Rechargeable Crimpers and Decappers with OLED Display Screen
Hundreds of operations from one charge
Supplier: Restek™ 23235
Description
- OLED screen display to optimize crimping power for each cap type, monitor battery use and view lifetime statistics
- Large battery allows for faster charge cycle and two sleep mode options extend battery life
- Easy to use
- Comfortable grip
- All kits include a universal plug set for battery charger
- For use with 11 mm and 20 mm caps
- One year warranty
Specifications
Crimper | |
20 mm | |
OLED display screen | |
1 year |
Caps | |
6.4 V | |
Charger USB drive and universal plug set with U.S., European, UK and Australian plug clips |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction