Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek Electronic Rechargeable Crimpers and Decappers with OLED Display Screen

Hundreds of operations from one charge

Manufacturer:  Restek 23238

 View more versions of this product

Catalog No. 06-611-691


Add to cart

Description

Description

  • OLED screen display to optimize crimping power for each cap type, monitor battery use and view lifetime statistics
  • Large battery allows for faster charge cycle and two sleep mode options extend battery life
  • Easy to use
  • Comfortable grip
  • All kits include a universal plug set for battery charger
  • For use with 11 mm and 20 mm caps
  • One year warranty
Specifications

Specifications

Caps
20 mm
Charger USB drive and universal plug set with U.S., European, UK and Australian plug clips
OLED display screen
6.4 V
Decapper
1 year
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit