Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Electronic Snap Circuits Mini Kits
Build six simple, everyday devices.
$13.38 - $20.20
Specifications
Min. Age | 8 |
---|---|
Min. Grade Level | 3 |
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
S90172
![]() |
Elenco Electronics Inc![]() SCP12 |
Electronic Snap Circuit Fm Radio | Tune it to your favorite fm station | 5.2 lb. | 2.35kg |
Each for $20.20
|
|
S90173
![]() |
Elenco Electronics Inc![]() SCP13 |
Electronic Snap Circuit Motion Detector | Movement triggers laser sounds | 24 lb. | 10.8kg |
Each for $20.20
|
|
S90176
![]() |
Elenco Electronics Inc![]() SCP06 |
Electronic Snap Circuit Flying Saucer | Press the switch to release the saucer into the air | 17 lb. | 7.71kg |
Each for $13.38
|
|
Description
- Build simple circuits to better understand how they work
- Easy-to-use kits simply snap together
- Grades 3+
Flying Saucer
- Press the switch to release the saucer into the air
- Tune it to your favorite FM station
- Movement triggers laser sounds
- Red light adds visual trigger to motion
- Records up to eight seconds
- Plays music after your recording
Specifications
8 | |
3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title