Learn More
Electrostatic Balance Kit Products for Science Education
Manufacturer:  Sempco Inc KT2002
Catalog No. S52094
Add to cart
Includes: Materials to construct 12 electrostatic balances. 6 Acetate strips, 6 pieces Woolen cloth, 30 Carbon coated balls, 12 Plastic vials, 50 Styrofoam cups, 100 pins, 24 insulating Silvers, straws to support the Carbon coated balls, 12 Plastic pivot supports.
Description
Description
Effects of static electricity and Coulomb's Law can be investigated with this inexpensive and easy-to-assemble electrostatic balance.
Provides:
  • Qualitative and quantitative data about static electric charges
  • How charges interact with each other
Specifications
Specifications
Education
Provides qualitative and quantitative data about static electric charges, how charges interact with each other
Materials to construct 12 electrostatic balances. 6 Acetate strips, 6 pieces Woolen cloth, 30 Carbon coated balls, 12 Plastic vials, 50 Styrofoam cups, 100 pins, 24 insulating Silvers, straws to support the Carbon coated balls, 12 Plastic pivot supports.
Electrostatic balance kit