Learn More
Market Lab Electrostatic Sprayer System
SDP
Manufacturer:  Market Lab ML23138
Encompass
With electrostatic technology that provides 360° coverage, you can trust the Protexus Electrostatic Sprayer to reach the areas standard disinfecting sprays miss. The efficient, easy-to-use sprayers allow for faster, safer, and more consistent application with a design that can cover a 450 sq ft room in just over two minutes and with less than a third of the disinfectant required by a hand-powered trigger sprayer.
Catalog No. NC1432363
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »