Learn More
Electrostatics Materials
Support individual electrostatic experiments for class size up to 24 students.
$18.80
Specifications
For Use With (Equipment) | Electroscope |
---|---|
For Use With (Application) | Experiments with electrified objects and electrostatic induction |
Includes:
Individual Kit:
- Two clear acetate strips and two colored vinyl strips; two low-density polyethylene insulating strips with wooden handles; six coated pith balls; two squares of wool cloth; two squares of cotton cloth; two aluminum tubes and nylon monofilament
Class Kit:
- Eight clear acetate strips; eight colored vinyl strips; eight low-density polyethylene insulating strips with wooden handles; 24 coated pith balls; eight squares of wool cloth; eight squares of cotton cloth; eight aluminum tubes and nylon monofilament
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S52093
![]() |
Sempco Inc
KT2003 |
Electrostatics materials individual kit |
Pack for $18.80
|
|
Description
Experiments with electrified objects and electrostatic induction can be performed easily with these kits. Choose between two different-sized kits small or large. An electroscope is recommended for each group (See S43262, S52057 or S43267).
Specifications
Electroscope | |
Experiments with electrified objects and electrostatic induction |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.