Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elements, Mixtures and Compounds Lab Investigation
Provides a basic understanding of the primary concepts of chemistry.
Specifications
For Use With (Application) | Provides a basic understanding of the primary concepts of chemistry |
---|---|
Includes | Individual software |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S63468
![]() |
Neo/Sci Corporation
201673 |
Elements, Mixtures And Compounds Lab Investigation Kit |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Investigate how matter is classified
- Identify various types of mixtures
- Compare the properties of solutions and mixtures
- Identify elements and compounds
- Study how chemical symbols, formulas and equations are used
- Class size: 40 students
Specifications
Provides a basic understanding of the primary concepts of chemistry | |
Individual software |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title