Learn More
Eletriptan hydrobromide, Tocris™
Orally active, selective 5-HT1B/1D agonist
Manufacturer:  R&D Systems 3862/10
Quantity 10mg
 View more versions of this product
Catalog No. 38-6210
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
463.43
Eletriptan hydrobromide
C22H26N2O2S·HBr
3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole hydrobromide
Specifications
177834-92-3
C22H26N2O2S·HBr
3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole hydrobromide
Eletriptan hydrobromide
>99%
463.43
10mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature