Learn More
ELF3 Mouse anti-Human, Clone: OTI9H7, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809987
Catalog No. 50-166-9830
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ESE-1 (Epithelium-Specific Ets) is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.
Specifications
Specifications
ELF3
Monoclonal
Unconjugated
ELF3
Lyophilized
ELF3
Human recombit protein fragment corresponding to amino acids 175-286 of human ELF3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI9H7
PBS with 8% trehalose and no preservative; pH 7.3
P78545
EPR-1, ERT, ESE-1, ESX
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1999