Learn More
ELF4 Mouse anti-Human, Clone: OTI4A3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810093
Catalog No. 50-166-9880
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5 ft.-WGGA-3 ft.. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications
Specifications
ELF4
Monoclonal
Unconjugated
ELF4
Lyophilized
ELF4
Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4A3
PBS with 8% trehalose and no preservative; pH 7.3
Q99607
ELFR, MEF
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2000