Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550955

Catalog No. PA550955


Add to cart

Description

Description

ELF4 Polyclonal Antibody detects endogenous levels of ELF4 protein.

ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5 ft.-WGGA-3 ft.. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications

Specifications

ELF4
Polyclonal
Unconjugated
ELF4
Liquid
ELF4
Synthesized peptide derived from the Internal region of human Elf-4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q99607, Q9Z2U4
ELFR, MEF
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2000, 56501
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit