Learn More
ELF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160201AP
Catalog No. 50-173-0649
Add to cart
Description
Description
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5 ft.-WGGA-3 ft.. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications
Specifications
ELF4
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99607, Q9Z2U4
E74 like factor 4, ELF4, ELFR, MEF, Myeloid Elf 1 like factor
Rabbit
IgG
150 μL
-20°C
Primary
2000, 302811, 56501
Immunoprecipitation, Western Blot
Unconjugated
ELF4
Liquid
Elf4
ELF4 Fusion Protein Ag8836
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat